Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Moticam® BTI10 Tablet Digital Camera, Motic

Supplier: Motic
Moticam® Motic Moticam
1100600101511 1100600101511
76595-464EA 1089.86 USD InStock
76595-464 CA76595-464
Moticam® BTI10 Tablet Digital Camera, Motic
Cameras
The Moticam BTI10 unlike other Moticams is combined with a 4.0MP and a 10" Android tablet that will deliver fast live images up to 30 fps. By using our pre-installed app, MotiConnect, simply log on with your tablet and begin to capture, edit, and save your images.

  • 10" LCD Android tablet with 1280×800 screen resolution
  • Connection: Wi-Fi (2.4 and 5 GHz), Ethernet
  • Wi-Fi resolution: 4.0 MP captured video resolution
  • Wi-Fi: Wi-Fi output (802.1 a/b/g/n/ac supporting both hot-spot mode or using existing Wi-Fi network)
  • Preloaded optimized MotiConnect app

Moticam® BTI is the most flexible tablet solution, consisting of a 4 MP camera with a built-in adjustable bracket that allows you to mount your own tablet. The camera can be connected via Wi-Fi to an iOS or android tablet. The Moticam BTI will help microscope users visualize, edit and share the microscope images, enabling computer-free digital microscopy.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty