Would you like to log in again?
TDE Series Ultra-Low Freezers with Chart Recorder, Thermo Scientific
Supplier: Thermo Scientific
TDE60086FARCR TDE50086FARCR TDE50086LDRCR TDE60086LARCR TDE60086LDRCR TDE50086FDRCR TDE60086FDRCR TDE50086LARCR
76595-616EA 26998.15 USD InStock
76595-616 76595-594 76595-642 76595-586 76595-578 76595-568 76595-634 76595-626
TDE Series Ultra-Low Freezers with Chart Recorder, Thermo Scientific
Freezers Ultra Low Temperature Freezers
TDE sereies ultra-low temperature freezers are part of the Standard Performance (STP) family.
Insulation: water-blown polyurethane foam
Certifications: cULus; FDA Class II.
Packaging: Pack includes one ULT freezer, two shelves of sliding drawer racks and boxes, and a factory-installed chart recorder.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty