Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Geno CL Laboratory Grade EDI Lab Water System, 1 to 15 MΩ-cm, Avidity Science

Supplier: Avidity Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

2311-5000-003 2311-5000-004 2311-5000-002 2311-5000-001 2311-5000-005
76595-738EA 62476.55 USD InStock
76595-738 76595-740 76595-742 76595-734 76595-736
Geno CL Laboratory Grade EDI Lab Water System, 1 to 15 MΩ-cm, Avidity Science
Water Purification Systems Deionizers
The Geno CL utilizes proven EDI water purification technologies to produce purified type 2 (DI) general laboratory grade water.

  • Flexibility and scalability provided by unique Electro deionization - Cassette Technology
  • Each water stream workings independently, ensuring no single point of failure
  • Cassettes can be independently replaced with minimal downtime and intervention
  • Data logging via secure link to protected cloud storage available
  • Remote diagnostics via an Ethernet or 4G connection available
  • Common fault output for connection to Building Management System available
  • 10" high resolution touchscreen
  • Intuitive, pictorial settings, reducing language barriers
  • Password protected interface
  • Alarm warnings with a two stage, time and date stamp
  • Consumable reminder with on screen video user guides
  • Can be used as single POU or Room distribution to multiple pieces of equipment/POU

The Geno CL water purification system provides a seamless supply of Type 2 (DI) water for the daily operations in laboratories. Featuring an intuitive touchscreen for ease of use and supported by our own service team, you can trust and rely on Geno CL to keep your equipment and testing running smoothly. Equipped with EDI water purification technology and loop distribution to provide the resilience required by modern laboratories without the need for multiple water purification units, saving valuable space within your laboratory.

Inorganics: >10 MΩ-cm
TOC content: <30 ppb
Silica: <0.05 mg/L
Particles: 0.2 (at µm filter)

Certifications: Manufactured to ISO 9001:2015 standard. Certified to IEC 61010-1:2010 and EN 61326-21:2013.

Delivery information: Ships within 8 to 10 weeks from receiving the PO.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty