Would you like to log in again?
Epredia™ 115 Laser Slotted Cassettes with Separate Lid
Supplier: Richard-Allan Scientific
Epredia™
115-01-10LM 115-11-10LM 115-04-10LM 115-09-10LM 115-12-10LM 115-03-10LM 115-08-10LM 115-07-10LM 115-05-10LM 115-10-10LM 115-02-10LM 115-06-10LM
76614-820CS 593.29 USD InStock
76614-820 76614-824 76614-812 76614-822 76614-810 77308-608 76614-816 76614-826 76614-814 76614-808 77308-606 76614-818
Epredia™ 115 Laser Slotted Cassettes with Separate Lid
Embedding Cassettes Cassettes for Labeling Instruments
Manufactured specifically for laser printing, the series 115 slotted cassettes come with separate lids and are available in 12 colors.
  • Series 115 laser cassettes are designed for use on both the Epredia™ NOVA™ and VEGA™ laser printers
  • They contain a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Comes in 12 different colors: white, yellow, blue, pink, green, orange, gray, aqua, lilac, beige, ochre and red with color-matched lids
  • Taped stacks of 40 cassettes; 25 stacks per case for a total of 1000 cassettes and lids
  • Series 115 laser cassette is not compatible with thermal tape style printers
Designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into the Epedia™ laser cassette hoppers for printing.
Ordering information: Lids included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty