Would you like to log in again?
Epredia™ 315 Laser Embedding Cassettes
Supplier: Richard-Allan Scientific
Epredia™
315-07-18LM 315-04-18LM 315-11-18LM 315-01-18LM 315-09-18LM 315-06-18LM 315-03-18LM FTML315 315-05-18LM 315-10-18LM 315-02-18LM 315-12-18LM 315-08-18LM
77308-640CS 857.64 USD InStock
77308-640 76614-860 77308-638 76614-864 76614-852 76614-862 76614-850 76614-856 76614-866 76614-854 76614-898 76614-848 76614-858
Epredia™ 315 Laser Embedding Cassettes
Embedding Cassettes Cassettes for Labeling Instruments
Manufactured specifically for laser printing, the series 315 embedding cassettes are available in 12 colors in this classical style of round holes.
  • Series 315 laser cassettes are designed for use on both the Epredia™ NOVA™ and VEGA™ laser printers
  • They contain a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Comes in 12 different colors: white, yellow, blue, pink, green, orange, gray, aqua, lilac, beige, ochre and red
  • Requires reusable metal stainless steel lids (sold separately)
  • Taped stacks of 40 cassettes and 45 stacks per case for a total of 1800 cassettes
  • Series 315 laser cassette is not compatible with thermal tape style printers
Designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into the Epredia™ laser cassette hoppers for printing.
Delivery information: Lids not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty