Would you like to log in again?
Panthera TEC EpiPolarizing Trinocular Microscope Packages, Motic
Supplier: Motic
MBPANPET-4000 MBPANPET-1080X MBPANPET-S6
76615-006EA 8173 USD InStock
76615-006 76615-008 76615-010
Panthera TEC EpiPolarizing Trinocular Microscope Packages, Motic
Microscopes
The panthera TEC POL models present a high flexibility for the inspection of all kind of polarizing materials, for transparent samples as well as for opaque specimen from technical education environments and industrial quality control.
  • Panthera TEC-EpiPolarizing trinocular microscope
  • 0.5× C-Mount camera adapter for ¹/₃" and ¹/₂" chip sensors
  • Motic analyzes basic 64-bit; USB + dongle (no charge)
  • Moticam S6/1080×/BMH4000
The compact Epi illuminator carries a slot for polarizer and analyzer, ready to set up polarization contrast and to reduce internal reflections. An intermediate tube with focusable/centerable bertrand lens is prepared for a conoscopic analyzis of crystals and minerals in transmitted light.
Ordering information: Contact your avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty