Would you like to log in again?
Moticam® S Series sCMOS Cameras, Motic
Supplier: Motic
Moticam®
MB-S1 MB-S6 MB-S3 MB-S12 MB-S20 MB-S6-MONO MB-PROS5L MB-PROS5PLUS
76615-024EA 1048.06 USD InStock
76615-024 76615-026 76615-018 76615-028 76615-030 76615-032 76615-020 76615-022
Moticam® S Series sCMOS Cameras, Motic
Cameras
Moticam® S series-high grade sCMOS cameras for an ultimate imaging result.
  • High-resolution USB cameras
  • Affordable with high-quality imaging
  • All-in-one-box with accessories and software included
  • Multi-platform software- runs on IOS, Android, Windows, Linux
Motics new sCMOS cameras, designed by German engineers and manufactured by professionals represent an impressive impetus to microscope camera performance. With a member of this product line the door for new application fields opens wide. The accelerated data readout enables to work on fast phenomena in biology and industry.
Delivery information: Package includes CS ring adaptor, USB 3.1 cable, Motic 4-dot calibration slide, Motic Images Plus 3.0 for PC/OSX/Linux.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty