Would you like to log in again?
VWR® RED Taq® DNA Polymerase Master Mix
Supplier: VWR
VWR® REDTaq®
V170306 V180308 V190306 V190303 V160306 V180307 V170303 V180303 V160303 V190309 V180309 V180306
76620-468EA 256.42 USD InStock
76620-468 76778-970 76620-478 76620-466 76620-476 76620-464 76620-474 76620-484 76620-472 76620-482 76620-470 76620-480
VWR® RED Taq® DNA Polymerase Master Mix
Nucleic Acid Reagents End-point PCR Enzymes and Kits
VWR® Red Taq DNA Polymerase Master Mix, which also contains an inert red dye, can be directly loaded onto an agarose gel without addition of electrophoresis loading buffers.
  • All-in-one master mix for great convenience
  • Timesaving reaction setup
  • Direct loading onto agarose gel
  • Easy visualisation of pipetting
  • Red tracking dye
  • Increased reproducibility
  • High product yield
  • Processes up to 5 kb
Red Taq DNA Polymerase master mixes are a good alternative to VWR® Taq DNA Polymerase. The inert red dye and stabilizer are extra features, which allow direct loading onto your agarose gel for analysis.
The Red Taq DNA Polymerase master mixes offer reduced set-up time and eliminate the risk of contamination of reaction components. Fewer handling steps lead to better reproducibility, which makes master mixes suitable for standard PCR applications, screening and high-throughput testing.
Red Taq DNA Polymerase Master Mix is a ready-to-use 1.1X or 2X reaction mix. Simply add primers, template, and water to carry out primer extensions and other molecular biology applications.
Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.
VWR® Red Taq DNA Polymerase 1.1X and 2X master mixes are available in two final MgCl2 concentrations: 1.5 mM and 2 mM.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty