Would you like to log in again?
VWR® GC Tempase Hot Start DNA Polymerase Master Mixes
Supplier: Avantor
VWR®
V227103 V331703 V331706 V332706
76620-524EA 363 USD InStock
76620-524 76620-500 76620-528 76620-526
VWR® GC Tempase Hot Start DNA Polymerase Master Mixes
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Ready to use GC TEMPase 2X Master Mixes I and II are designed for amplification of GC-rich sequences.
  • High success rate with amplification of GC-rich DNA
  • High sensitivity, specificity and product yield
  • Diminished formation of non-specific products
  • Reaction set-up at room temperature
  • Processes up to 4 kb
The GC TEMPase 2X master mixes consist of TEMPase Hot Start DNA Polymerase combined with an optimized GC buffer system (GC buffer I or II), dNTPs and MgCl2. Just add DNA template and primers to carry out PCR.
GC-Rich DNA Target Kit consists of TEMPase Hot Start DNA polymerase and seperate tubes with GC buffer I and II. The kit is designed for initial testing before using one of the GC-TEMPase 2X Master Mixes.
Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty