Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Betaine enhancer solution 5M

Supplier: VWR International
Synonyms: TrimethylglycineTMG

V351104
76620-530EA 64.37 USD InStock
76620-530
Betaine enhancer solution 5M
Betaine
Betaine enhancer is especially effective when used with high GC-rich regions or templates with a high degree of secondary structures. It has a decreasing effect on the primer melting temperature.

  • Enhances amplification on GC-rich and difficult DNA targets
  • Lowers the melting temperature

The performance of Betaine Enhancer Solution 5 M is superior compared to standard enhancers such as formamide, DMSO, TMAC, BSA and non-ionic detergents. Betaine Enhancer Solution is an excellent enhancer especially when used with GC-rich regions or templates with a high degree of secondary structures.

Betaine has a decreasing effect on the melting temperature of DNA and primers. Therefore, denaturation temperatures as well as primer annealing temperatures should be reduced by 1 – 5 °C. The optimal annealing temperature should be determined individually for each reaction.

Caution: For Research Use Only. Not for use in diagnostic procedures.


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00012123
CAS Number: 107-43-7
Order Now
Specification Test Results
Functional test - 1X Betaine Enchancer Solution is tested for amplification of 613 bp fragment from human genomic DNA using Taq DNA polymerase at standard PCR Conditions Passes test
Volume check- fill volume >= 0.1 ml per tube Passes test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty