Would you like to log in again?
VWR® TAQ Plus DNA Polymerase Master Mix
Supplier: VWR
VWR®
V370506 V370503 V370501 V370507
76620-534EA 59.22 USD InStock
76620-534 76620-540 76620-538 76620-536
VWR® TAQ Plus DNA Polymerase Master Mix
Nucleic Acid Reagents End-point PCR Enzymes and Kits
VWR® Taq Plus is an optimized format of Taq DNA polymerase master mix and, therefore, is a good alternative to Taq DNA polymerase and Taq DNA polymerase master mix.
  • Ready to use 2X master mix for greater convenience
  • Significantly reduced set-up time
  • Increased specificity compared to Taq DNA polymerase master mix
  • Reduced risk of contamination
  • Processes up to 4 kb
Ensures increased specificity and improved PCR performance, even on difficult templates. These reagents are suitable for standard PCR applications, screening and high-throughput testing.
This PCR master mix is composed of Taq DNA polymerase and an optimized buffer system, including dNTPs and magnesium chloride (1.5 mM).
Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty