Would you like to log in again?
VWR® dNTPs
Supplier: VWR
VWR®
V511104 V502004 V511107 V500005
76620-546EA 160.43 USD InStock
76620-546 76620-544 76620-542 76620-548
VWR® dNTPs
Nucleic Acid Reagents Nucleotides
Ready to use molecular biology grade dNTP mixes and dNTP sets.
  • Available as pre-mixed 40 or 100 mM solutions , or as sets of individual 100 mM dNTP solutions
  • Both pre-mixed and sets have been functionally tested in PCR
  • Purity >99% by HPLC
  • Supplied in solution at pH 7.3 to 7.5
The dNTP mix is designed to save hands-on time for researchers, and reduce the possibility of contamination by reducing pipetting. The dNTP mix and dNTP solutions are convenient for use in DNA polymerization reactions, DNA labelling and sequencing processes.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty