Would you like to log in again?
VWR® DNA Loading Buffers
Supplier: Avantor
VWR®
V608104 V608204 V608304
76620-554EA 40.3 USD InStock
76620-554 76620-552 76620-550
VWR® DNA Loading Buffers
Stains Electrophoresis Stains
VWR® loading buffers, with three different dyes and migration fronts, make it easy to select the right loading buffer for your specific task.
  • Ready-to-use buffers
  • 5x formulation
  • Three different tracking dyes available
VWR® loading buffers are formulated as 5x solutions containing Ficoll, Tris-buffer, EDTA and either Cresol Red, Bromophenol Blue or Orange G as tracking dye.
DNA loading buffers are used for loading DNA samples onto DNA gels for gel electrophoresis. DNA loading buffers contain a coloured dye and a density agent. The density agent serves to enhance the density of the DNA sample allowing the DNA to sink into the bottom of the well. The dye adds visibility to the DNA sample and also serves as a tracking dye allowing the user to monitor the DNA migration during electrophoresis.
For long term storage, keep the loading buffer at -20 °C.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty