Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® DNA Ladders

Supplier: Avantor
VWR®
V610341 V610241 V610141
76620-556EA 277.52 USD InStock
76620-556 76620-560 76620-558
VWR® DNA Ladders
Electrophoresis Reagents Molecular Weight Markers
DNA ladders are supplied in a loading buffer that is ready to use on agarose and polyacrylamide DNA gels. The ladders are suitable with TBE, TAE, SB and LB electrophoresis systems.

  • Supplied in ready to use loading buffer
  • For direct loading and easy visualisation
  • Clear, distinct bands
  • Mass-calibrated bands for DNA quantification
  • PCR DNA LADDER has a 1000 bp band that is extra bright to serve as reference point
  • Blue dye front running at 100 to 300 bp at 0.5 to 1.5% agarose

High Range DNA Ladder molecular range: 200 to 12000 bp; mass-calibrated bands from 6 to 40 ng/band for DNA quantification.

Low Range DNA Ladder molecular range: 100 to 1000 bp; mass-calibrated bands from 8 to 40 ng/band for DNA quantification.

PCR DNA Ladder molecular range: 100 to 3000 bp; mass-calibrated bands from 10 to 30 ng/band for DNA quantification.

High Range DNA Ladder, Low Range DNA Ladder and PCR DNA Ladder are supplied in 0.5 mL packs sufficient for 250 lanes.

Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty