Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® DNA Ladders

Supplier: Avantor
VWR®
V610641 V610541 V610441
76620-566EA 59.22 USD InStock
76620-566 76620-564 76620-562
VWR® DNA Ladders
Electrophoresis Reagents Molecular Weight Markers
DNA ladders are supplied in a loading buffer that is ready to use on agarose and polyacrylamide DNA gels. The ladders are suitable with TBE, TAE, SB and LB electrophoresis systems.

  • Supplied in ready to use loading buffer
  • For direct loading and easy visualisation
  • Clear, distinct bands
  • Blue dye front running at 100 to 300 bp at 0.5 to 1.5% agarose

Mini DNA Ladder molecular range: 100 to 500 bp

Low DNA Ladder molecular range: 100 to 1000 bp

PCR Ladder molecular range: 100 to 3000 bp

Mini DNA Ladder, Low DNA Ladder and PCR Ladder are supplied in 0.5 mL packs for 100 lanes.

Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty