Would you like to log in again?
VWR® ExoCleanUp FAST
Supplier: Avantor
VWR® ExoCleanUp FAST
V620607 V620601 V620603
76620-568EA 109.89 USD InStock
76620-568 76620-572 76620-570
VWR® ExoCleanUp FAST
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
ExoCleanUp FAST PCR reagent is a one-step PCR clean-up reagent for optimal sequencing results, consisting of a balanced combination of a heat-labile exonuclease I (HL-ExoI) and a recombinant shrimp alkaline phosphatase (rSAP).
  • Designed to clean-up PCR products in 5 minutes
  • No need for spin columns or magnetic beads
  • Treatment improves downstream applications, such as DNA sequencing and SNP analysis
Treatment of amplified PCR products with this reagent helps to remove residual primers and single-stranded DNA, and inactivates excess dNTPs by dephosphorylation. After enzymatic treatment at 37 °C for a minimum of 2 minutes, this reagent is completely inactivated by heating at 80 °C for a minimum of 3 minutes.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty