Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Fast HiFi DNA Polymerase

Supplier: VWR
VWR®
V767501 V767506 V767503 V767504
76620-578EA 907.39 USD InStock
76620-578 76620-576 76620-574 76620-580
VWR® Fast HiFi DNA Polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Fast HiFi DNA Polymerase (2 U/µL) is a proofreading DNA polymerase with the following features: high fidelity >60X Taq DNA Polymerase, ability to amplify problematic DNA targets, such as those with low to high GC content, and ability to perform amplification on long DNA targets.

  • High fidelity: >60X Taq Fidelity
  • 3’→5’ proofreading exonuclease activity
  • Processes up to 18 kb (human gDNA) and 25 kb (λDNA)
  • Good coverage on DNA templates with low to high GC content
  • Fast extension rate: 10 s/kb

Fast HiFi DNA Polymerase: 2 Units/μl

Fast HiFi DNA Polymerase exhibits both 5'→3' DNA polymerase activity and 3'→5' proofreading exonuclease activity enabling this polymerase to correct base pair mismatches. It is supported with 5X Fast HiFi Buffer allowing robust amplification on DNA targets with low to high GC content and long DNA targets.

For difficult amplicons, such as GC-rich DNA samples, those with complex secondary structures or long amplicons, the addition of 1 to 2 M Betaine Enhancer Solution is recommended.

Fast HiFi DNA Polymerase is recommended for applications which require extremely high fidelity, such as cloning/subcloning, NGS applications, and mutagenesis.

Sample types: gDNA, plasmid DNA, bacterial DNA, λDNA and other DNA templates.

Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.

For more convenient handling, Fast HiFi DNA Polymerase is also available as a 2X master mix.

Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty