Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Fast HiFi DNA Polymerase 2X Master Mix

Supplier: VWR
VWR®
V770101 V770103 V770106 V770107
76620-588EA 7015.95 USD InStock
76620-588 76620-586 76620-584 76620-582
VWR® Fast HiFi DNA Polymerase 2X Master Mix
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Fast HiFi DNA Polymerase Master Mix is a ready-to-use 2X reaction mix composed of Fast HiFi DNA polymerase and an optimized buffer system, including dNTPs and magnesium chloride, allowing robust amplification on DNA target with low to high GC content and long DNA targets.

  • All-in-one 2X master mix for great convenience
  • High fidelity: >60X Taq Fidelity
  • 3’→5’ proofreading exonuclease activity
  • Processes up to 11 kb from complex templates
  • Good coverage on DNA templates with low to high GC content
  • Fast extension rate: 10 s/kb

Fast HiFi DNA Polymerase 2X Master Mix is recommended for applications which require extremely high fidelity, such as cloning /subcloning, NGS applications and mutagenesis.

Sample types: gDNA, plasmid DNA, bacterial DNA, λDNA and other DNA templates.

The Fast HiFi DNA Polymerase exhibits both 5'→3' DNA polymerase activity and 3'→5' proofreading exonuclease activity enabling this polymerase to correct base pair mismatches.

For difficult amplicons, such as GC-rich DNA samples, those with complex secondary structures or long amplicons, the addition of 1 to 2 M Betaine Enhancer Solution is recommended.

Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.

Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty