Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
chemiSOLO Imager, Azure Biosystems
Supplier: Azure Biosystems
CS1000 CS1002 CS1001 CS1003
76628-048EA 13711.5 USD InStock
76628-048 76628-038 76628-070 76628-040
chemiSOLO Imager, Azure Biosystems
Photodocumentation Systems
The chemiSOLO is a personal Western blot imager that offers high-quality chemiluminescent and visible imaging using simple and easy workflows, all within a compact unit.
  • Femtogram detection of chemiluminescent Westerns
  • Visible imaging for protein gel densitometric analysis
  • Controllable by computer, mobile device, OR tablet, which enables flexibility and accessibility of use
  • Compact design fits neatly into busy lab spaces
  • Applications for chemiluminescence, visible protein gels, densitometry
The chemiSOLO’s advanced imaging technology, intuitive controls, and compact design provide users with a personal, high-quality chemiluminescent imager that will fit neatly into their busy lab spaces. It provides sensitive chemiluminescent imaging with femtogram protein detection capability and up to 24-bit imaging to capture images with increased dynamic range for more quantitative Western blots. The flexibility of controlling the imager using a wide range of devices, including tablets and mobile devices, through a unique browser-based system allows the chemiSOLO to effortlessly bring digital chemiluminescent imaging to every lab.
Certifications: In compliance with CE RED, CB scheme, cTUVus, and FCC standards.
Delivery information: Supplied with an Azure Test Target, USB with user manual and backup files, power cable, warranty registration card, and installation card.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty