Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
CompactDry™ Pseudomonas aeruginosa (PA), Hardy Diagnostics
Supplier: Hardy Diagnostics
CompactDry™
PA100 PA100
76628-082PK 329.48 USD InStock
76628-082 CA76628-082
CompactDry™ Pseudomonas aeruginosa (PA), Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
CompactDry™ Pseudomonas aeruginosa (PA) is a ready-to-use test method recommended for the enumeration of Pseudomonas aeruginosa in foods, beverages, water, cannabis, and more.
  • Sturdy frame, stackable packaging
  • Self-diffusing media
  • Ability to subculture without damage to colonies
  • No leakage
  • Easy to label
CompactDry™ PA is suitable for testing a broad range of water types for human consumption or other related industries.
Each cassette comes with four connected plates that can be easily snapped apart to support a variety of dilution scenarios.
Caution: Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty