Would you like to log in again?
SMS Sequential Sterilization Wraps, Blue, Heavy Duty, 50 gsm, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KCRP-300-0404 KCRP-300-1515 KCRP-300-0202 KCRP-300-4040 KCRP-300-0909 KCRP-300-0808 KCRP-300-2424 KCRP-300-0303 KCRP-300-4548 KCRP-300-01.5X01.5 KCRP-300-5454 KCRP-300-3030 KCRP-300-0606 KCRP-300-1212 KCRP-300-4848 KCRP-300-3636 KCRP-300-1818 KCRP-300-6060 KCRP-300-1822 KCRP-300-2020 KCRP-300-6080 KCRP-300-0912 KCRP-300-5472 KCRP-300-4545
76628-176CS 501.6 USD InStock
76628-176 76628-088 76628-132 76628-144 76628-156 76628-178 76628-146 76628-102 76628-136 76628-158 76628-138 76628-116 76628-108 76628-090 76628-160 76628-094 76628-150 76628-162 76628-140 76628-086 76628-152 76628-120 76628-164 76628-098
SMS Sequential Sterilization Wraps, Blue, Heavy Duty, 50 gsm, Keystone Cleanroom Products
Sterilization Wraps
Keystone Cleanroom Products' line of heavy-duty SMS wrappers provide an excellent microbial barrier. Ideal for pharmaceutical production applications.
  • Manufactured and packaged in a certified ISO 8 Cleanroom
  • Clean - no cellulose
  • Strong, rip and tear resistant 50 gsm SMS material
  • An extensive range of sizes available means less cutting
  • Hydrophobic barrier allows sterilization gases, but prevents liquid and microbial penetration
  • Sub packed into sealed labeled poly bags with lot number and date of manufacture for complete traceability
For use in steam, ethylene oxide and hydrogen peroxide sterilization processes.
Certifications: Certificates of Conformance sent with shipments.
Packaging: The SMS wraps are packed in sealed, labeled poly bags with lot number and expiration date for traceability.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty