Would you like to log in again?
SMS Sequential Sterilization Wraps, Blue, Super Duty, 88 gsm, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KCRP-600-0808 KCRP-600-4848 KCRP-600-1515 KCRP-600-0303 KCRP-600-1822 KCRP-600-6060 KCRP-600-2020 KCRP-600-4548 KCRP-600-0912 KCRP-600-5472 KCRP-600-0202 KCRP-600-5454 KCRP-600-3636 KCRP-600-2424 KCRP-600-01.5X01.5 KCRP-600-4040 KCRP-600-1212 KCRP-600-0404 KCRP-600-1818 KCRP-600-6080 KCRP-600-0606 KCRP-600-4545 KCRP-600-3030 KCRP-600-0909
76628-110CS 564.3 USD InStock
76628-110 76628-154 76628-100 76628-166 76628-122 76628-112 76628-134 76628-168 76628-124 76628-114 76628-104 76628-126 76628-148 76628-128 76628-106 76628-118 76628-092 76628-170 76628-084 76628-172 76628-096 76628-130 76628-174 76628-142
SMS Sequential Sterilization Wraps, Blue, Super Duty, 88 gsm, Keystone Cleanroom Products
Sterilization Wraps
Ideal for pharmaceutical and biotech production applications.
  • Manufactured and packaged in a certified ISO 8 cleanroom
  • Clean - no cellulose
  • Strong, rip and tear resistant 88 gsm SMS material
  • An extensive range of sizes available means less cutting
  • Hydrophobic barrier allows sterilisation gases, but prevents liquid and microbial penetration
  • Sub packed into sealed labelled poly bags with lot number and date of manufacture for complete traceability
For use in steam, ethylene oxide and hydrogen peroxide sterilization processes.
Certifications: Certificates of Conformance sent with shipments.
Packaging: The SMS wraps are packed in sealed, labeled poly bags with lot number and expiration date for traceability.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty