The Allegra V-15R benchtop centrifuges bring more versatility to the laboratory bench. These compact, refrigerated centrifuges are ideal for evolving laboratories.
- Free up precious benchtop space
- Support multiple workflows with just one instrument
- Advanced temperature management
- Wide choice of centrifuge packages available to suit various applications
The Allegra V-15R centrifuges maintain a consistent temperature, allowing you to spin faster and longer without overheating your samples. All while Rapid Temp and Eco Mode allow for even more efficiency and functionality.
The Allegra V-15R centrifuges are designed to be a workhorse in the laboratory. Operation is simple and seamless thanks to an easy-to-use interface and ergonomic design. With 10 rotor options at your fingertips, you will be ready and able to support multiple workflows with just one instrument.
10 rotors and a comprehensive selection of adapters allows for a variety of applications, from cell and blood separation to high throughput screening. Its compact footprint frees up space on your bench and its ≤55 dBA (at fullspeed) makes it a quiet companion in the lab.
Operation is simple and seamless with soft-touch lid closure, 50 programmable runs and an easy-to-use illuminated interface.