Would you like to log in again?
Allegra V-15R Benchtop Centrifuges, Beckman Coulter
Supplier: Beckman Coulter
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Allegra®
C63381 C63444 C63432 C63421 C63386 C63438 C69864 C63429 C63125 C69867 C63380 C63124 C63387 C63423 C63435 C63445 C69866 C63125 C63383 C63420 C63380 C63445 C69865 C63426 C63444 C63439 C63381 C63375 C63432 C63124 C63427 C69879 C69878 C63375 C63433 C63433 C63450 C63447 C63389 C63441 C63450 C63377
76628-198EA 18188.5 USD InStock
76628-198 76628-210 76521-710 76521-732 76521-730 76521-708 76521-706 76521-728 76521-726 76521-704 76521-702 76521-724 CA76521-732 CA76521-734 76628-184 76628-186 76628-188 76628-200 76628-202 76628-204 CA76521-730 76521-722 76628-206 76521-700 76521-720 76628-208 CA76521-708 76521-718 76521-716 76521-714 76521-712 76521-734 76628-190 CA76521-722 CA76521-700 76628-192 CA76521-702 CA76521-724 76628-194 CA76521-704 76628-196 CA76521-706
Allegra V-15R Benchtop Centrifuges, Beckman Coulter
Centrifuges Benchtop Centrifuges
The Allegra V-15R benchtop centrifuges bring more versatility to the laboratory bench. These compact, refrigerated centrifuges are ideal for evolving laboratories.
  • Free up precious benchtop space
  • Support multiple workflows with just one instrument
  • Advanced temperature management
  • Wide choice of centrifuge packages available to suit various applications
The Allegra V-15R centrifuges maintain a consistent temperature, allowing you to spin faster and longer without overheating your samples. All while Rapid Temp and Eco Mode allow for even more efficiency and functionality.
The Allegra V-15R centrifuges are designed to be a workhorse in the laboratory. Operation is simple and seamless thanks to an easy-to-use interface and ergonomic design. With 10 rotor options at your fingertips, you will be ready and able to support multiple workflows with just one instrument.
10 rotors and a comprehensive selection of adapters allows for a variety of applications, from cell and blood separation to high throughput screening. Its compact footprint frees up space on your bench and its ≤55 dBA (at fullspeed) makes it a quiet companion in the lab.
Operation is simple and seamless with soft-touch lid closure, 50 programmable runs and an easy-to-use illuminated interface.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty