Would you like to log in again?
Electro-Gel™ Electrolyte Replenishers
Supplier: Bio-Serv
EGS-1 EGS16P-1 EGS16P-1 EGS-1
76628-300PK 34.68 USD InStock
76628-300 CA76628-300 CA76406-464 76406-464
Electro-Gel™ Electrolyte Replenishers
Animal Supplies Animal Feed
Gelled, oral electrolyte hydration supplement. Provides balanced electrolyte and glucose levels to provide energy, nutrients and hydration to accelerate recovery.
  • Economical and easy to use requiring no refrigeration or special storage
  • Highly palatable with a sweet, salty orange or strawberry flavor
  • Comparable to Pedialyte and PRANG in gel form
  • Suitable for a variety of research species
  • Thermoreversible
  • Gamma Irradiated
Shelf life of 12 months, cool dry storage.
Ingredients: Purified water, dextrose, gelatin, L-ascorbic acid phosphate, sodium chloride, sodium bicarbonate, citric acid, potassium chloride, flavoring, natural color, potassium sorbate, acesulfame potassium.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty