Would you like to log in again?
Nutra-Gel™ Complete Nutrition
Supplier: Bio-Serv
Nutra-Gel™
NGB8P-2 NGB-1 NGB-2 NGB-2 NGB-1 NGB8P-2
76406-484CS 148.38 USD InStock
76406-484 76628-354 CA76628-354 CA76406-486 76406-486 CA76406-484
Nutra-Gel™ Complete Nutrition
Animal Supplies Animal Feed
Gelled, 100% highly palatable, nutritionally complete purified (AIN 93G), bacon flavored diet.
  • Provides both food and water to rodents
  • Economical and easy to use requiring no refrigeration or special storage Phytoestrogen free
  • Bacon flavor stimulates appetite
  • Thermoreversible
  • Nutritionally assayed
  • Gamma irradiated
Use for chewing and mobility impairments, recovering and debilitated rodents, weanlings and transportation.
Shelf life of six months, cool dry storage.
Ingrediants: Purified Water, Sucrose, Casein, Gelatin, Corn Starch, Maltodextrin, Soybean Oil, Cellulose, Mineral Mix (AIN-93G), Bacon Flavor, Vitamin Mix (AIN-93), L-Cystine, Choline Bitartrate, Xanthan Gum, Vitamin E Acetate, Sorbic Acid, tBHQ.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty