Would you like to log in again?
Difco™ TC Yeastolate, Ultra-Filtered (UF), Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
Difco™
292804 292805 670079 670079 292804 292805
76628-770EA 28406.7 USD InStock
76628-770 76628-750 CA76628-770 CA76628-750 76628-836 CA76628-836
Difco™ TC Yeastolate, Ultra-Filtered (UF), Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Difco TC Yeastolate, ultra-filtered (UF), is an animal origin-free (AOF) product derived from water-soluble portions of autolyzed yeast (Saccharomyces cerevisiae). This product is intended as a nutritional supplement for bacterial, insect, and mammalian cell culture.
  • Mixture of low molecular weight peptides, amino acids, carbohydrates (simple and complex), as well as vitamins
  • Meets additional requirements for reduced endotoxin, clarity, and suitability for the biopharmaceutical industry
  • Ultra-filtered
  • Supplementation of TC yeastolate to CHO cell culture can enhance growth and mAb production
These products are intended as nutritional supplements for bacterial, insect, and mammalian cell culture. TC yeastolate has been used in insect cell culture. TC yeastolate was found to be a very versatile supplement to enhance growth and production characteristics of Sf9 and Gibco™ High Five™ cells [1-5]. Additionally, supplementation of TC yeastolate to CHO cell culture can enhance growth and mAb production.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty