Would you like to log in again?
Difco™ Soytone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
Difco™
212488 212489 212489 212488
76628-744EA 137.21 USD InStock
76628-744 76628-756 CA76628-744 CA76628-756
Difco™ Soytone, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Difco Soytone is an enzymatic digest of soy. With high concentrations of vitamins and carbohydrates uniquely present in soy, this supplement offers an alternative carbohydrate profile to yeast supplements for mammalian cell processes, such as CHO.
  • High concentrations of vitamins and carbohydrates
  • An enzymatic digest of soy
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty