Would you like to log in again?
Difco™ Phytone™ Supplement, Ultra-Filtered (UF), Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
Difco™
210936 210936 210931 210931
76628-760EA 2686.99 USD InStock
76628-760 CA76628-760 76628-728 CA76628-728
Difco™ Phytone™ Supplement, Ultra-Filtered (UF), Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Difco Phytone, ultra-filtered (UF), is an animal origin-free enzymatic digest of soy. Due to the naturally high concentrations of vitamins and carbohydrates in soy, Difco Phytone UF is a nutritious and excellent carbohydrate source.
  • Ultra-filtered
  • Animal origin-free nutritional supplement
  • Excellent carbohydrate source due to the naturally high concentrations of vitamins and carbohydrates in soy
  • Desirable for use with mammalian cell culture applications where reduced endotoxin levels are important
  • Recommended for use in media for the cultivation of a wide variety of organisms, including fungi
This product is an ultra-filtered version of Phytone peptone, making it desirable for use with mammalian cell culture applications where reduced endotoxin levels are important.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty