Would you like to log in again?
Bacto™ Brain Heart Infusion, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Bacto™
237300 237300
76628-774EA 2942.73 USD InStock
76628-774 CA76628-774
Bacto™ Brain Heart Infusion, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Prepared Media for Microbiology
Bacto Brain Heart Infusion is a general-purpose medium used to cultivate aerobic and anaerobic bacteria from a variety of clinical and nonclinical materials.
  • Can be used in tests for the verification of fecal streptococci
  • Listed by the National Committee for Clinical Laboratory Standards (NCCLS) as a medium for use in the preparation of microdilution trays for antimicrobial susceptibility testing by the broth microdilution procedure
Bacto Brain Heart Infusion is used in the cultivation of fastidious and non-fastidious microorganisms, including bacteria (Streptococci, Pneumococci, and Meningococci), yeasts, and molds. 
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty