Would you like to log in again?
Resurge™ CD1 Supplement, without Glucose and L-Glutamine, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Resurge™
670011 670011 670013 670013 670012 670012
76628-778EA 227.94 USD InStock
76628-778 76628-856 CA76628-856 CA76628-778 76628-948 CA76628-948
Resurge™ CD1 Supplement, without Glucose and L-Glutamine, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Resurge CD1 Supplement, without glucose and L-glutamine, is a chemically defined supplement formulation to complement a variety of base media.
  • Used in CHO mammalian culture systems
  • Can be used either as a media supplement or a culture system feed
This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Resurge CD1 Supplement, without glucose and L-glutamine, was developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty