Would you like to log in again?
Bacto™ Neopeptone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Bacto™
211681 211680 211681 211680
76628-796EA 7983.36 USD InStock
76628-796 76628-820 CA76628-796 CA76628-820
Bacto™ Neopeptone, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Bacto Neopeptone is an enzymatic digest of bovine (milk) and porcine protein. This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation.
  • Enzymatic digest of bovine (milk) and porcine protein
  • Animal origin nutritional supplement
  • Contains a wide variety of peptide sizes in combination with vitamins, nucleotides, and minerals
  • Recommended for use in media for detection of fungi
  • Reported to provide nutrients for support of Spirochetes and Protozoa
  • Microbial fermentation
  • Production of vaccines for animal health applications
Customers have used Bacto Neopeptone as a component of culture media in the cultivation of human pathogens, notably Bordetella pertussis and group A Streptococci, as well as Borrelia burgdorferi and Clostridium species, to produce vaccines for animal health applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty