Would you like to log in again?
OneFeed™ Supplement, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
OneFeed™
670109 670108 670110 670108 670110 670109
76628-872EA 1535.44 USD InStock
76628-872 CA76628-872 76628-798 CA76628-798 76628-846 CA76628-846
OneFeed™ Supplement, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Gibco™ OneFeed™ Supplement is a versatile chemically defined, animal origin–free, protein-free feed that can be used with a variety of media to enhance protein.
  • Chemically defined (CD) and animal origin-free (AOF)
  • Developed using several different cell lines in various culture systems with a variety of commercially available media
  • Balanced nutritional profile, providing key components to support high growth and production throughout the culture life
  • Can be used a feed supplement to enhance growth and production of a variety of CHO lines
This high-performance feed has been developed using several different cell lines in various culture systems with a variety of commercially available media. OneFeed Supplement has a balanced nutritional profile that provides key components to support high growth and production throughout the culture life.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty