Would you like to log in again?
Bacto™ TC Yeastolate, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
Bacto™
292731 255771 255772 292731 255772 255771
76628-792EA 76.55 USD InStock
76628-792 CA76628-792 76628-810 CA76628-810 76628-802 CA76628-802
Bacto™ TC Yeastolate, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Bacto TC Yeastolate is an animal origin-free hydrolysate from the water-soluble portion of autolyzed yeast. This supplement contains a mixture of peptides, amino acids, carbohydrates (simple and complex), nucleosides, and vitamins.
  • Mixture of low molecular weight peptides, amino acids, carbohydrates (simple and complex), as well as vitamins
  • Meets additional requirements for reduced endotoxin, clarity, and suitability for the biopharmaceutical industry
  • Bacterial, insect, and mammalian cell culture
  • Supplementation of TC yeastolate to CHO cell culture can enhance growth and mAb production
Customers have used this supplement for CHO-based production of biotherapeutic monoclonal antibodies used in oncological and rheumatological therapeutics and recombinant proteins. Additionally, customers have used this supplement in combination with a soy supplement, such as Difco Phytone, ultra-filtered (UF), to provide an optimal titer performance in mammalian cell based processes by providing additional vitamins and carbohydrates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty