Would you like to log in again?
Resurge™ CD Pak, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Resurge™
670030 670030
76628-818EA 725.76 USD InStock
CA76628-818 76628-818
Resurge™ CD Pak, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Resurge CD Pak are a diverse set of chemically defined supplement formulations to complement a variety of base media. The CD PAK contains one of each Resurge supplement: CD1, CD2, CD3, CD4, and CD5 at 100 grams each.
  • The Resurge CD Pak contains the following supplements:
  • Resurge CD1 Supplement without glucose and L-glutamine
  • Resurge CD2 Supplement without glucose and L-glutamine
  • Resurge CD3 Supplement without glucose and L-glutamine
  • Resurge CD4 Supplement without glucose and L-glutamine
  • Resurge CD5 Supplement without glucose and L-glutamine
The supplements have been developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. In order to increase versatility of the formulation, glucose and L-glutamine are not included in the final formulations. Therefore, these two components should be added by users and monitored as appropriate.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty