Would you like to log in again?
Bacto™ Casamino Acids, Technical, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
Bacto™
223110 223120 223110 223120
76628-808EA 131.54 USD InStock
76628-808 CA76628-808 76628-824 CA76628-824
Bacto™ Casamino Acids, Technical, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Bacto Casamino Acids, technical, is an animal origin (AO) acid hydrolysis of casein. In contrast to the Bacto Casamino Acids, this supplement has been processed less and, as a result, has higher sodium chloride and iron content.
  • An animal origin (AO) acid hydrolysis of casein
  • Less processing results in higher sodium chloride and iron content
  • Used in the production of animal health vaccines
  • Supports the growth of Bordetella, Haemophilus, and Vibrio species
Bacto Casamino Acids, Technical provides similar benefits to Bacto Casamino Acids, for applications requiring a less refined hydrolysate.
Certifications: AOAC, BAM, COMPF, SMWW, USDA, USP, ISO/DIS, 11014.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty