Would you like to log in again?
Difco™ Beef Extract, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Difco™
212610 212610
76628-838EA 238.14 USD InStock
76628-838 CA76628-838
Difco™ Beef Extract, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Difco Beef extract is an nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins.
  • Derived from infusion of beef
  • Provides a rich source of nutrients
  • Available as a paste Intended to replace aqueous infusion of meat in microbiological culture media
Provided as a paste, customers have used Difco Beef extract in a variety of microbial fermentation cell lines, including Escherichia coli, Bacillus subtillis, Pasteurella multocida, and Streptococcus species to produce numerous animal health vaccines.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty