Would you like to log in again?
Acidicase™ Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Acidicase™
211843 211843
76628-844EA 147.43 USD InStock
76628-844 CA76628-844
Acidicase™ Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Acidicase Peptone is a hydrochloric acid hydrolysate of casein. The manufacturing process produces a casein hydrolysate, which has a high salt content of approximately 37% and nitrogen content of approximately 8%.
  • Nutritional supplement in vitamin assay, susceptibility testing and other laboratory media and microbial fermentation
The hydrolysis of the casein, a milk protein rich in amino acid nitrogen, is carried out until all the nitrogen is converted to amino acids or other compounds of relative simplicity. Casein contains little cystine and is deficient in tryptophan, which is destroyed by the acid treatment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty