Would you like to log in again?
BiTek™ Proteose Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
BiTek™
253310 253310
76628-848EA 2630.87 USD InStock
76628-848 CA76628-848
BiTek™ Proteose Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
BiTek Proteose Peptone is an enzymatic digest of protein, developed as an alternative to Bacto Proteose Peptones for scale-up to production applications.
  • similar growth characteristics
  • Used in preparing microbiological culture media
  • Used in producing bacterial toxins
Bacto Proteose Peptones are enzymatic digests of protein. Studies of peptic digests of animal tissue prepared under varying digestion parameters demonstrated that no single peptone is the most suitable nitrogen source for every microbiological application. These findings led to the development of Bacto Proteose Peptone, Bacto Proteose Peptone No. 2, and Bacto Proteose Peptone No. 3. Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty