Would you like to log in again?
Difco™ Malt Extract Broth, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Difco™
214912 214912
76628-854EA 2233.92 USD InStock
76628-854 CA76628-854
Difco™ Malt Extract Broth, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Prepared Media for Microbiology
Difco™ Malt Extract Broth is a complete microbiological media for the cultivation of yeasts and molds. Difco™ Malt Extract Broth contains malt extract, which provides the carbon, nitrogen, and nutrient sources required for growth of microorganisms.
  • Contains malt extract
  • Recommended for yeasts and molds in the U.S. Food and Drug Administration's Bacteriological Analytical Manual
The broth also has a high concentration of carbohydrates, such as maltose and dextrose, as an energy source required for fermentation by yeasts and molds. Yeast extract provides additional nitrogen, carbon, vitamins, and cofactors required for growth. The acidic pH of Difco™ Malt Extract Broth promotes optimal growth of yeasts and molds while restricting bacterial growth.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty