Would you like to log in again?
Difco™ Casamino Acids, Vitamin Assay, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
Difco™
228820 228820 228830 228830
76628-860EA 134.95 USD InStock
76628-860 CA76628-860 76628-954 CA76628-954
Difco™ Casamino Acids, Vitamin Assay, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Difco Casamino Acids, vitamin assay, is an animal origin (AO) acid hydrolysis of casein having low sodium chloride and iron concentrations.
  • Prepared according to the method described by Mueller and Miller
  • Supplemented with growth factors, cystine, maltose and iron and inorganic salts
  • Successful in fermentation for Pneumococcal, Diptheria, Pertussis, Tetanus and Zoster vaccines
  • Supports the growth of Bordetella, Corynebacterium, Pseudomonas, Haemophilus and Escherichia
Difco Casamino Acids, vitamin assay, is an animal origin (AO) acid hydrolysis of casein having low sodium chloride and iron concentrations. It is an excellent supplement for media formulations where nitrogen requirements are minimal, including diphtheria toxin production.
Certifications: AOAC, BAM, COMPF, SMWW, USDA, USP, ISO/DIS, 11014.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty