Would you like to log in again?
Bacto™ TC Lactalbumin Hydrolysate, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
Bacto™
259962 259961 259961 259962
76628-862EA 2268 USD InStock
76628-862 CA76628-862 76628-758 CA76628-758
Bacto™ TC Lactalbumin Hydrolysate, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Bacto TC Lactalbumin Hydrolysate, an animal origin peptone, is the enzymatically hydrolyzed protein portion of milk Whey.
  • Animal origin (AO) peptone
  • Mixture of peptides, amino acids, and carbohydrates (simple and complex)
  • Can be used in mammalian, bacterial and insect cultures
  • Used in recombinant proteins applications
  • Used in animal health vaccines
Bacto TC Lactalbumin Hydrolysate, an animal origin peptone, is the enzymatically hydrolyzed protein portion of milk whey. This supplement is a mixture of peptides, amino acids, and carbohydrates, both simple and complex. Customers have successfully used Bacto TC Lactalbumin Hydrolysate to support the growth of many mammalian, bacterial, and insect cell types, and it has been used to produce both recombinant proteins and animal health vaccines. Customers have also used this supplement as an amino acid supplement for mammalian cell culture media.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty