Would you like to log in again?
Bacto™ Malt Extract, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Bacto™
218630 218610 218630 218610
76628-864EA 53.85 USD InStock
76628-864 CA76628-864 76628-958 CA76628-958
Bacto™ Malt Extract, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Bacto Malt Extract is the water-soluble portion of malted barley.
  • High concentration of reduced sugars, especially the maltoses, makes this suitable for the culture of yeasts and molds
  • Malt extract in the agar form is recommended for the detection and isolation of yeasts and molds from dairy products and foods, and as a medium for stock culture maintenance
Due to the extraction process, this animal origin-free supplement provides a very high carbohydrate content to any medium. Bacto Malt Extract has been used successfully in microbial fermentation applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty