Would you like to log in again?
Biosate™ Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Biosate™
294312 211862 294312 211862
76628-794EA 202.99 USD InStock
76628-794 76628-874 CA76628-794 CA76628-874
Biosate™ Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Biosate Peptone is an animal origin (AO) mixed hydrolysate comprised of 65% pancreatic digest of casein and 35% yeast extract. This combination provides synergistic nutritional benefits that are not provided by the components alone, resulting in a unique nutritional profile.
  • Hydrochloric acid hydrolysate of casein
  • Animal origin nutritional supplement
  • Milk protein rich in amino acid nitrogen
Intended for use as a nutritional supplement in vitamin assays, susceptibility testing, and other laboratory media, and microbial fermentation in which the high salt content will not interfere.
Customers have shown the combined use of these two peptones, when supplied as Biosate Peptone, improves toxin production with Clostridial species for vaccines, such as Clostridium botulinum and Clostridium perfringens.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty