Would you like to log in again?
Beef Extract Powder, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
212303 212303
76628-876EA 231.34 USD InStock
76628-876 CA76628-876
Beef Extract Powder, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Beef Extract Powder is a meat extract dried into a powder. This nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins. Additionally, due to differences in the process, beef extract powder has a different molecular weight profile than other beef extract products in our portfolio. 
  • Derived from infusion of beef
  • Provides a rich source of nutrients
  • Free-flowing, homogeneous powder
  • Applications - Intended to replace aqueous infusion of meat in microbiological culture media
Customers have used Beef Extract Powder in a variety of microbial fermentation cell lines including Clostridium sp., Escherichia coli, Bacillus subtillis, Pasteurella multocida, and Streptococcus species to produce numerous animal health vaccines.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty