Would you like to log in again?
Resurge™ CD3 Supplement, without Glucose and L-Glutamine, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
Resurge™
670019 670019 670020 670018 670020 670018
76628-884EA 1948.22 USD InStock
76628-884 76628-940 CA76628-940 CA76628-884 76628-816 CA76628-816
Resurge™ CD3 Supplement, without Glucose and L-Glutamine, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Resurge CD3 Supplement, without glucose and L-glutamine, is an animal origin-free (AOF) protein-free chemically defined supplement. It can be used in batch and fed batch mode in cell culture processes.
  • Due to the balanced nutritional nature of this supplement, rich in amino acids, it has also been used in bacterial applications
  • Can be used in batch and fed batch mode in cell culture processes
  • Use in bacterial applications
Resurge CD3 Supplement is formulated without glucose, L-glutamine, and sodium bicarbonate. Due to the balanced nutritional nature of this supplement, rich in amino acids, it has also been used in bacterial applications. All components are chemically defined with known molecular formulae and structures and meet our stringent Animal Origin-Free (AOF) specification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty