Would you like to log in again?
Starter Pak No. 1, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
215366 215366
76628-894PK 331.13 USD InStock
76628-894 CA76628-894
Starter Pak No. 1, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Used in animal and human health applications, Starter Pak No. 1 features a combination of yeast- and soy-based peptones. Three of the products in this pack have been ultra-filtered to reduce endotoxin levels. The yeast products add a mixture of peptides, amino acids, carbohydrates, nucleosides, and vitamins.
Provides conveniently packaged, 100-gram, non-GMP samples of these peptones that are best suited for use in human and animal health applications and contains:
Difco TC Yeastolate UF and Bacto TC Yeastolate - these peptones are ideal for CHO-based applications of biotherapeutic monoclonal antibodies and recombinant proteins.
Difco Yeast Extract UF and Bacto Yeast Extract, Technical - these peptones support optimal growth of many microbial species for a variety of human and animal health vaccines.
Difco Phytone Supplement, UF - this enzymatic digest of soy is an excellent, nutritious source of carbohydrates and is used in mammalian cell culture. This peptone works well alone and when blended with yeast-based peptones.
The Starter Paks provide conveniently packaged samples of the peptones most commonly used in mammalian cell culture and microbial fermentation bioproduction processes. Starter Paks are tailored for specific applications, including the production of monoclonal antibodies, recombinant proteins, and vaccines.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty