Would you like to log in again?
Bacto™ Tryptose, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
Bacto™
211713 211709 211709 211713
76628-952EA 278.96 USD InStock
76628-952 76628-898 CA76628-898 CA76628-952
Bacto™ Tryptose, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Bacto Tryptose is a mixed enzymatic hydrolysate with distinctive nutritional properties. The digestive process of tryptose results in assorted peptides of higher molecular weight suitable for long-chain amino acid requirements.
  • Reported as beneficial for cell culture applications
  • Animal origin (AO) nutritional supplement
  • Tryptose is very useful for cultivation of Streptococci, Pneumococci, Meningococci, and other Fastidious organisms
  • Tryptose Peptone is often used as a biomass enhancer for recombinant Escherichia coli production
For use in cell culture, Bacto Tryptose is a mixed enzymatic hydrolysate with distinctive nutritional properties. The digestive process of tryptose results in assorted peptides of higher molecular weight suitable for long-chain amino acid requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty