Would you like to log in again?
Bacto™ Casamino Acids, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Bacto™
223030 223020 223020 223030 223050 223050
76628-910EA 421.9 USD InStock
76628-910 76628-944 CA76628-944 CA76628-910 76628-904 CA76628-904
Bacto™ Casamino Acids, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Bacto Casamino Acids is an animal origin (AO) acid hydrolysis of casein. This supplement contains low salt and iron content making it an excellent supplement for media formulations where nitrogen requirements are minimal.
  • An animal origin (AO) acid hydrolysis of casein
  • Contains low salt and iron content, making it an excellent supplement for media formulations where nitrogen requirements are minimal
  • Maintains many of the nutritional benefits of hydrolyzed casein, but is further supplemented with growth factors, cystine, maltose, and an optimum amount of iron and inorganic salts
  • Supports the growth of Bordetella, Corynebacterium, Pseudomonas, Haemophilus, and Escherichia species
  • Successfully used in fermentation applications for Pneumococcal, Diptheria, Pertussis, Tetanus and Zoster vaccines
  • Together with Bacto Yeast Extract, technical, this supplement has been successfully used in the production of animal and human health vaccines
It maintains many of the nutritional benefits of hydrolyzed casein, but is further supplemented with growth factors, cystine, maltose, and an optimum amount of iron and inorganic salts.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty