Would you like to log in again?
Polypeptone™ Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Polypeptone™
211910 297108 297108 211910
76628-916EA 190.51 USD InStock
CA76628-906 CA76628-916 76628-916 76628-906
Polypeptone™ Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Polypeptone Peptone is a mixture of peptones made up of equal parts of pancreatic digest of casein and peptic digest of animal tissue.
  • Animal origin mixture of peptones
  • Includes the high content of amino acids
Found to meet nutritional requirements of various bacteria, fungi, and mammalian cells for which a single source of casein or meat peptones has been unsatisfactory.
Polypeptone Peptone includes the high content of amino acids and small polypeptides characteristic of pancreatic digest of casein and the larger polypeptides characteristic of peptic digest of animal tissue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty